Learn More
Eppendorf™ Refrigerated Multipurpose Centrifuge 5804 R Rectangular Bucket Adapters
Bucket adapters for use with Eppendorf 5804R Centrifuge.
Supplier:  Eppendorf™ 022637614
 View more versions of this product
Catalog No. 36-100-7189
Only null left
Explore available promotions
Description
Description
For use with Standard and Vacutainer conical tubes and micro test tubes.
Specifications
Specifications
Bucket Adapter
Rectangular Bucket (100 mL)
2
Standard and Vacutainer conical tubes and micro test tubes
Conical
17.5 mm
15 mL
Product Suggestions
Product Suggestions