Learn More
Eppendorf™ Refrigerated Multipurpose Centrifuge 5804 R Rectangular Bucket Adapters
Bucket adapters for use with Eppendorf 5804R Centrifuge.
Supplier: Eppendorf™ 022638327
Description
For use with Standard and Vacutainer conical tubes and micro test tubes.
Specifications
| Bucket Adapter | |
| 20 mm | |
| 7 to 18 mL |
| Rectangular Bucket (250 mL) | |
| 8 | |
| Standard and Vacutainer conical tubes and micro test tubes |