missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Reagent Reservoir
Description
- Filling volume 50–60mL
- One set = 10 pieces
Specifications
Specifications
| Capacity (Metric) | 60 mL |
| Material | Polypropylene |
| Sterility | Non-sterile |
| Type | Reagent Reservoir |
| Autoclavable | Autoclavable |