Learn More
Eppendorf™ Pipette Pick-a-Pack Sets
Catalog No. 14285904
Change view
Click to view available options
Volume (Metric):
10, 20, 200, 1000 μL
2.5, 10, 100, 1000 μL
2.5, 20, 200, 1000 μL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
14285904 10, 20, 200, 1000 μL
05403151 2.5, 10, 100, 1000 μL
05403152 2.5, 20, 200, 1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14285904 Supplier Eppendorf™ Supplier No. 2231300006
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
4-Pack sets available with Eppendorf Research™ plus single-channel pipettes in a variety of volume ranges.
Research Plus Features:
  • Includes four pipetters and one full rack of Eppendorf pipet tips for each pipetter volume
Specifications
Autoclavable Autoclavable
Number of Channels 1
Product Line Research Plus
Type Pipette Package
Volume (Metric) 10, 20, 200, 1000 μL
Includes Pipet 4-pack (0.5-10 μL, 2-20 μL yellow, 20-200 μL, 100-1,000 μL variable-volume pipettes) and epT.I.P.S. pipette tips.