missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Mastercycler™ Nexus X2 Thermal Cyclers
Provide simple, flexible programming with choice of gradient or nongradient blocks
$16,623.73 - $23,473.70
| Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
|
E6336000023
|
Eppendorf™
6336000023 |
Mastercycler Nexus GX2; With Gradient |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
E6339000024
|
Eppendorf™
6339000024 |
Mastercycler Nexus X2e; Without Gradient |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
E6338000020
|
Eppendorf™
6338000020 |
Mastercycler Nexus GX2e; With Gradient |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
E6337000027
|
Eppendorf™
6337000027 |
Mastercycler Nexus X2; Without Gradient |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- Able to run two totally independent protocols at same time
- Two different blocks for smaller (32-well) and larger (64-well) assays
- Larger block features optional (GX2/GX2e) gradient function for PCR optimization
- Can be combined with Eppendorf's PCR Tubes, PCR Strips or divisible plates
- Intuitive graphic programming
- Universal block for strips, 0.1 mL, 0.2 mL and 0.5 mL PCR tubes
- Small footprint and E-mail notification
- flexlid concept allows use of all types of consumables with automatic height adjustment of lid
- Heating technology of the block: Peltier Elements, Triple Circuit Technology
- Interfaces : USB, Ethernet, CAN in/out
- Universal Aluminum thermal block
Mastercycler nexus X2/X2e
- Without gradient
- Lid temperature range: 37 to 100°C
- With gradient
- Gradient block: Over 12 columns
- Gradient range: 1 to 20°C
- Gradient temperature range: 30 to 99°C
Specifications
| 120 V, 50/60 Hz 700 W |