Learn More
Eppendorf™ HeatSealer S100 Accessory, Plate Adapters
For use with HeatSealer S100
Supplier:  Eppendorf™ 5391070018
 View more versions of this product
Catalog No. 05412613
Only null left
Description
Description
  • Hermetic sealing of multiwell plates
  • Eliminates evaporation in PCR, reducing cross contamination
  • Seals 96- and 384-well plates
  • Integrated thermostat prevents overheating
  • Improved mechanics for even easier sealing
  • PCR Plate Adapter included
Warranty
One year
Specifications
Chemical Identifiers
320 W
HeatSealer S100
170°C
Specifications
320 W
HeatSealer S100
170°C
Low Profile Adapter
Product Suggestions
Product Suggestions