Learn More
Eppendorf™ HeatSealer S100 Accessory, Plate Adapters
Catalog No. 05412612
Change view
Click to view available options
Product Type:
High Profile Adapter
Low Profile Adapter
PCR Plate Adapter
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
05412612 High Profile Adapter
05412613 Low Profile Adapter
05412614 PCR Plate Adapter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 05412612 Supplier Eppendorf™ Supplier No. 5391070034
For use with HeatSealer S100
  • Hermetic sealing of multiwell plates
  • Eliminates evaporation in PCR, reducing cross contamination
  • Seals 96- and 384-well plates
  • Integrated thermostat prevents overheating
  • Improved mechanics for even easier sealing
  • PCR Plate Adapter included
Warranty
One year
Specifications
Wattage 320 W
For Use With (Equipment) HeatSealer S100
Sealing Temperature Range 170°C
Product Type High Profile Adapter