missing translation for 'onlineSavingsMsg'
Learn More

Epoxidized soya bean oil, MilliporeSigma™ Supelco™

Catalog No. 111018534
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018534 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018534 Supplier Merck Emd Millipore Supplier No. 439562.5GF

Epoxidized soya bean oil may be used as a reference standard for the accurate detection of regulated dyes. These dyes, although banned in many countries, are still being used illicitly as additives in food products.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 8013-07-8
Density 0.997 g/mL
Flash Point 231°C
Physical Form Neat
MDL Number MFCD00163560
Synonym ESBO; Soybean oil, epoxidized
Type Analytical Standard
Quantity 5 g
Grade Analytical Standard