Learn More
Agilent BioTek Epoch 2 Microplate Spectrophotometer
GREENER_CHOICE
Catalog No. BTEPOCH2TS
Change view
Click to view available options
Model:
EPOCH2NS
EPOCH2NSC
EPOCH2TS
EPOCH2TSC
Includes:
Cuvette Port
No Touchscreen, No Cuvette Port
Touchscreen only
Touchscreen; Cuvette port
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Includes
BTEPOCH2TS EPOCH2TS Touchscreen only
BTEPOCH2NS EPOCH2NS No Touchscreen, No Cuvette Port
BTEPOCH2NSC EPOCH2NSC Cuvette Port
BTEPOCH2TSC EPOCH2TSC Touchscreen; Cuvette port
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. BTEPOCH2TS Supplier Agilent BioTek Supplier No. EPOCH2TSSN
Get a Microvolume Plate with Purchase
View all promotions
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The BioTek Epoch 2 microplate spectrophotometer delivers excellent performance for UV-Vis absorbance measurements. For Research Use Only. Not for use in diagnostic procedures.
The BioTek Epoch 2 microplate spectrophotometer delivers excellent performance for UV-Vis absorbance measurements. Measurements can be performed in 6- to 384-well microplates, cuvets, and in microvolume samples with the available Take3 microvolume plate. The broad wavelength range enables applications from nucleic acid and protein quantification in the low UV to microbial growth assays higher wavelengths
Epoch 2 is easy to use and the optional touch screen interface makes it simple to choose predefined protocols or to define custom programs. Endpoint, kinetic, spectral scanning and well area scanning modes, plus incubation and shaking enable wide-ranging applications.
Specifications
Detector Type Photodiode
Light Source Xenon Flash Lamp
Measurement Speed 96 wells: 8 sec., 384 wells: 14 sec.
Accuracy ±2 nm
Certifications/Compliance CE and TUV marked. RoHS compliant.
For Use With (Application) Nucleic acid and protein quantification to ELISA and microbial growth assays
Height (English) 22 in.
Includes Touchscreen only
Model EPOCH2TS
Wavelength Range 200 to 999 nm, 1 nm increments
Frequency 50/60 Hz
Show More Show Less