missing translation for 'onlineSavingsMsg'
Learn More

Epithiochlorohydrine, 97%, Thermo Scientific™

Chemical Identifiers

CAS 3221-15-6
Molecular Formula C3H5ClS
Molecular Weight (g/mol) 108.59
MDL Number MFCD00041199
InChI Key XRWMHJJHPQTTLQ-UHFFFAOYSA-N
Synonym 2-chloromethyl thiirane, thioepichlorohydrin, epithiochlorohydrin, thioepichlorhydrin, chloropropylene sulfide, chloromethyl thiirane, 3-chloropropylene sulfide, 2-thiiranylmethyl chloride, thiirane, chloromethyl, chloropropylene sulphide
PubChem CID 18588
IUPAC Name 2-(chloromethyl)thiirane
SMILES C1C(S1)CCl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC291390050
SDS View Documents
Thermo Scientific Chemicals
291390050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC291390250
SDS View Documents
Thermo Scientific Chemicals
291390250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

3221-15-6
108.59
XRWMHJJHPQTTLQ-UHFFFAOYSA-N
18588
C1C(S1)CCl
C3H5ClS
MFCD00041199
2-chloromethyl thiirane, thioepichlorohydrin, epithiochlorohydrin, thioepichlorhydrin, chloropropylene sulfide, chloromethyl thiirane, 3-chloropropylene sulfide, 2-thiiranylmethyl chloride, thiirane, chloromethyl, chloropropylene sulphide
2-(chloromethyl)thiirane

Specifications

3221-15-6
100.0
69°C to 70°C (67.0mmHg)
96% min. (GC)
C3H5ClS
MFCD00041199
1.275
Solubility in water: .. Other solubilities: soluble in methanol,chloroform,diethylether
C1C(S1)CCl
108.59
108.59
Epithiochlorohydrine
96.0
1.2750g/mL
Authentic
Glass bottle
1.5240 to 1.528
5g
2-chloromethyl thiirane, thioepichlorohydrin, epithiochlorohydrin, thioepichlorhydrin, chloropropylene sulfide, chloromethyl thiirane, 3-chloropropylene sulfide, 2-thiiranylmethyl chloride, thiirane, chloromethyl, chloropropylene sulphide
XRWMHJJHPQTTLQ-UHFFFAOYSA-N
2-(chloromethyl)thiirane
18588
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Causes serious eye damage.
Fatal if swallowed.
May cause an allergic skin reaction.
Toxic in contact with skin.
Toxic if inhaled.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Danger

SDS
Documents

Documents

RUO – Research Use Only