missing translation for 'onlineSavingsMsg'
Learn More

Epithiochlorohydrine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 291390050

 View more versions of this product

Catalog No. AC291390050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Epithiochlorohydrine
96.0
1.2750g/mL
Authentic
Glass bottle
1.5240 to 1.528
5g
2-chloromethyl thiirane, thioepichlorohydrin, epithiochlorohydrin, thioepichlorhydrin, chloropropylene sulfide, chloromethyl thiirane, 3-chloropropylene sulfide, 2-thiiranylmethyl chloride, thiirane, chloromethyl, chloropropylene sulphide
XRWMHJJHPQTTLQ-UHFFFAOYSA-N
2-(chloromethyl)thiirane
18588
97%
3221-15-6
100.0
69°C to 70°C (67.0mmHg)
96% min. (GC)
C3H5ClS
MFCD00041199
1.275
Solubility in water: .. Other solubilities: soluble in methanol,chloroform,diethylether
C1C(S1)CCl
108.59
108.59

Chemical Identifiers

3221-15-6
108.59
XRWMHJJHPQTTLQ-UHFFFAOYSA-N
18588
C1C(S1)CCl
C3H5ClS
MFCD00041199
2-chloromethyl thiirane, thioepichlorohydrin, epithiochlorohydrin, thioepichlorhydrin, chloropropylene sulfide, chloromethyl thiirane, 3-chloropropylene sulfide, 2-thiiranylmethyl chloride, thiirane, chloromethyl, chloropropylene sulphide
2-(chloromethyl)thiirane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Causes serious eye damage.
Fatal if swallowed.
May cause an allergic skin reaction.
Toxic in contact with skin.
Toxic if inhaled.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only