missing translation for 'onlineSavingsMsg'
Learn More

Epirubicin hydrochloride

Catalog No. AAJ60411MA
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60411MA 10 mg
AAJ60411MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60411MA Supplier Thermo Scientific Chemicals Supplier No. J60411MA

Cell-permeable anthracycline anti-tumor antibiotic | CAS: 56390-09-1 | C27H30ClNO11 | 579.983 g/mol

Cell-permeable anthracycline anti-tumor antibiotic

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56390-09-1
Molecular Formula C27H30ClNO11
Molecular Weight (g/mol) 579.983
MDL Number MFCD00941448
InChI Key MWWSFMDVAYGXBV-DPZLWMTJSA-N
Synonym Epiadriamycin
PubChem CID 123134690
IUPAC Name (7S,9S)-7-[(2R,4R,5R)-4-amino-5-hydroxy-6-methyloxan-2-yl]oxy-6,9,11-trihydroxy-9-(2-hydroxyacetyl)-4-methoxy-8,10-dihydro-7H-tetracene-5,12-dione;hydrochloride
SMILES CC1C(C(CC(O1)OC2CC(CC3=C(C4=C(C(=C23)O)C(=O)C5=C(C4=O)C=CC=C5OC)O)(C(=O)CO)O)N)O.Cl

Specifications

Melting Point 185°C
Color Red
Quantity 10 mg
Merck Index 14,3623
Solubility Information Soluble in DMSO to 100mM, or in ethanol to 10mM
Formula Weight 579.98
Physical Form Solid
Chemical Name or Material Epirubicin hydrochloride
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 260-145-2
RTECSNumber QI9295750
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only