CAS RN 56390-09-1
IUPAC Name:
hydrogen (8S,10S)-10-{[(2R,4S,5R,6S)-4-amino-5-hydroxy-6-methyloxan-2-yl]oxy}-6,8,11-trihydroxy-8-(2-hydroxyacetyl)-1-methoxy-5,7,8,9,10,12-hexahydrotetracene-5,12-dione chloride
Synonyms:
hydrogen epirubicin chloride
Molecular Weight (g/mol):
579.98
Molecular Formula:
C27H30ClNO11
InChi Key:
MWWSFMDVAYGXBV-FGBSZODSSA-N
SMILES:
[H+].[Cl-].COC1=CC=CC2=C1C(=O)C1=C(O)C3=C(C[C@](O)(C[C@@H]3O[C@H]3C[C@H](N)[C@@H](O)[C@H](C)O3)C(=O)CO)C(O)=C1C2=O
1
–
1
of
1
results
1
Epirubicin hydrochloride
CAS: 56390-09-1 Molecular Formula: C27H30ClNO11 Molecular Weight (g/mol): 579.983 MDL Number: MFCD00941448 InChI Key: MWWSFMDVAYGXBV-DPZLWMTJSA-N Synonym: Epiadriamycin PubChem CID: 123134690 IUPAC Name: (7S,9S)-7-[(2R,4R,5R)-4-amino-5-hydroxy-6-methyloxan-2-yl]oxy-6,9,11-trihydroxy-9-(2-hydroxyacetyl)-4-methoxy-8,10-dihydro-7H-tetracene-5,12-dione;hydrochloride SMILES: CC1C(C(CC(O1)OC2CC(CC3=C(C4=C(C(=C23)O)C(=O)C5=C(C4=O)C=CC=C5OC)O)(C(=O)CO)O)N)O.Cl