missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse Epiregulin (EREG) ELISA Kit

ELISA

$916.00

Specifications

Accession Number Q61521
Assay Range 8.23 to 6000 pg/mL
Assay Sensitivity 10 pg/mL
Calibration Range 10 pg/mL to 6000 pg/mL
Assay Solid-phase sandwich ELISA
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
EMEREG
Encompass
View Documents Promotion Details
Invitrogen™
EMEREG
96 Tests
Pack for $916.00
Only null left
 
Description

Description

The Mouse Epiregulin (Ms EREG) ELISA quantitates Ms Epiregulin in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms Epiregulin. Principle of the method The Mouse Epiregulin solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Epiregulin is an EGF-related growth factor that binds specifically to EGFR (ErbB1) and ErbB4, but not ErbB2 or ErbB3. It is expressed mainly in the placenta and peripheral blood leukocytes, as well as in certain carcinomas of the bladder, lung, kidney and colon. Epiregulin stimulates the proliferation of keratinocytes, hepatocytes, fibroblasts and vascular smooth muscle cells. It also inhibits the growth of several tumor-derived epithelial cell lines. Human Epiregulin is initially synthesized as a glycosylated 19.0 kDa transmembrane precursor protein, which is processed by proteolytic cleavage to produce a 6.0 kDa mature secreted sequence.
Specifications

Specifications

Q61521
10 pg/mL
Solid-phase sandwich ELISA
ELISA
Plasma, Serum, Supernatant
Mouse
Colorimetric Microplate Reader
Ereg
<10
HRP
Angiogenesis
2°C to 8°C
Epiregulin
4 hrs 45 mins
8.23 to 6000 pg/mL
10 pg/mL to 6000 pg/mL
HRP
Growth Factors & Receptors
This ELISA kit shows no cross-reactivity with the following cytokines tested: Mouse CD30, L CD30, T CD40, CRG-2, CTACK, CXCL16, Eotaxin , Eotaxin-2, Fas Ligand, Fractalkine, GCSF, GM-CFS, IFN- gamma, IGFBP-3, IGFBP-5, IGFBP-6, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-3 Rb, IL-4, IL-5, IL-9, IL-10, IL-12 p40/p70, IL-12 p70, IL-13, IL-17, KC, Leptin R, LEPTIN(OB), LIX, L-Selectin, Lymphotactin, MCP-1, MCP- 5, M-CSF, MIG, MIP-1 alpha, MIP-1 gamma, MIP-2, MIP-3 beta, MIP-3 alpha, PF-4, PSelectin, RANTES, SCF, SDF-1 alpha, TARC, TCA-3, TECK, TIMP-1, TNF-alpha, TNF RI, TNF RII, TPO, VCAM-1, VEGF.
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
13874
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
RUO
Plasma 50 μL, Serum 50 μL, Supernatant 100 μL
EPR
1 hrs 20 mins
Videos
SDS
Documents

Documents