missing translation for 'onlineSavingsMsg'
Learn More

Epinephrine Bitartrate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111019242
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019242 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019242 Supplier Merck Emd Millipore Supplier No. PHR1509500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Epinephrine belongs to the class of drugs termed as catecholamines, which exerts its action on both alpha and beta-adrenergic receptors using a G protein-linked second messenger system. Featured Industries: Pharmaceutical (small molecule)

Specifications

Packaging Ampule
CAS 51-42-3
Linear Formula C9H13 NO3 · C4H6O6
MDL Number MFCD00035077
Synonym (-)-Epinephrine (+)-bitartrate salt; L-3,4-Dihydroxy-alpha-(methylaminomethyl)benzyl alcohol D-hydrogen bitartrate salt; L-Adrenaline (+)-bitartrate salt; L-Adrenaline D-hydrogentartrate; L-Epinephrine D-hydrogentartrate
Type Certified Reference Material
Quantity 500 mg
Formula Weight 333.29
Grade Certified Reference Material