missing translation for 'onlineSavingsMsg'
Learn More

Epinastine Hydrochloride 98.0+%, TCI America™

Catalog No. E07991G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E07991G 1 g
E0799100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E07991G Supplier TCI America Supplier No. E07991G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 108929-04-0
Molecular Formula C16H16ClN3
Molecular Weight (g/mol) 285.78
MDL Number MFCD00933434
InChI Key VKXSGUIOOQPGAF-UHFFFAOYNA-N
Synonym 3-Amino-9,13b-dihydro-1H-dibenzo[c,f]imidazo[1,5-a]azepine Hydrochloride
PubChem CID 157313
ChEBI CHEBI:51037
IUPAC Name hydrogen 2,4-diazatetracyclo[12.4.0.0²,⁶.0⁷,¹²]octadeca-1(18),3,7,9,11,14,16-heptaen-3-amine chloride
SMILES [H+].[Cl-].NC1=NCC2N1C1=CC=CC=C1CC1=CC=CC=C21

Specifications

Melting Point 275°C
Color White
Quantity 1 g
Formula Weight 285.78
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Epinastine Hydrochloride
TSCA No
Recommended Storage Refrigerator