accessibility menu, dialog, popup

UserName

CAS RN 108929-04-0

IUPAC Name: hydrogen 2,4-diazatetracyclo[12.4.0.0²,⁶.0⁷,¹²]octadeca-1(18),3,7,9,11,14,16-heptaen-3-amine chloride
Synonyms: epinastine hydrochloride
Molecular Weight (g/mol): 285.78
Molecular Formula: C16H16ClN3
InChi Key: VKXSGUIOOQPGAF-UHFFFAOYNA-N
SMILES: [H+].[Cl-].NC1=NCC2N1C1=CC=CC=C1CC1=CC=CC=C21
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results