missing translation for 'onlineSavingsMsg'
Learn More

(-)-Epigallocatechin gallate, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111016012
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111016012 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111016012 Supplier Merck Emd Millipore Supplier No. 9389410MG

(−)-Epigallocatechin gallate is a bioactive compound, which is an important constituent of green tea. It may serve as a potent candidate to prevent cancer.

Featured Industries: Food and Beverages

Specifications

CAS 989-51-5
Physical Form Neat
Linear Formula C22H18O11
MDL Number MFCD00075940
Synonym (-)-cis-2-(3,4,5-Trihydroxyphenyl)-3,4-dihydro-1(2 H)-benzopyran-3,5,7-triol 3-gallate; (-)-cis-3,3′,4′,5,5′,7-Hexahydroxy-flavane-3-gallate; EGCG
Type Analytical Standard
Quantity 10 mg
Formula Weight 458.37
Percent Purity ≥98% (HPLC)
Grade Analytical Standard