missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(-)-Epicatechin Gallate 98.0+%, TCI America™
$498.62 - $1251.36
Chemical Identifiers
| CAS | 1257-08-5 |
|---|---|
| Molecular Formula | C22H18O10 |
| Molecular Weight (g/mol) | 442.376 |
| MDL Number | MFCD00075936 |
| InChI Key | LSHVYAFMTMFKBA-TZIWHRDSSA-N |
| Synonym | =--epicatechin gallate, epicatechin gallate, =--epicatechin-3-o-gallate, l-epicatechin gallate, =--epicatechin-3-gallate, =--epicatechingallate, epicatechin monogallate, --epicatechin 3-o-gallate, teatannin, epicatechol, gallate |
| PubChem CID | 107905 |
| ChEBI | CHEBI:70255 |
| IUPAC Name | [(2R,3R)-2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-3,4-dihydro-2H-chromen-3-yl] 3,4,5-trihydroxybenzoate |
| SMILES | C1C(C(OC2=CC(=CC(=C21)O)O)C3=CC(=C(C=C3)O)O)OC(=O)C4=CC(=C(C(=C4)O)O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
E089020MG
|
TCI America
E089020MG |
20 mg |
Each for $498.62
|
|
|||||
|
E0890100MG
|
TCI America
E0890100MG |
100 mg |
Each for $1,251.36
|
|
|||||
Chemical Identifiers
| 1257-08-5 | |
| 442.376 | |
| LSHVYAFMTMFKBA-TZIWHRDSSA-N | |
| 107905 | |
| [(2R,3R)-2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-3,4-dihydro-2H-chromen-3-yl] 3,4,5-trihydroxybenzoate |
| C22H18O10 | |
| MFCD00075936 | |
| =--epicatechin gallate, epicatechin gallate, =--epicatechin-3-o-gallate, l-epicatechin gallate, =--epicatechin-3-gallate, =--epicatechingallate, epicatechin monogallate, --epicatechin 3-o-gallate, teatannin, epicatechol, gallate | |
| CHEBI:70255 | |
| C1C(C(OC2=CC(=CC(=C21)O)O)C3=CC(=C(C=C3)O)O)OC(=O)C4=CC(=C(C(=C4)O)O)O |
Specifications
| 1257-08-5 | |
| C22H18O10 | |
| 20 mg | |
| LSHVYAFMTMFKBA-TZIWHRDSSA-N | |
| [(2R,3R)-2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-3,4-dihydro-2H-chromen-3-yl] 3,4,5-trihydroxybenzoate | |
| 107905 | |
| 442.38 | |
| Crystalline Powder |
| 255°C | |
| MFCD00075936 | |
| =--epicatechin gallate, epicatechin gallate, =--epicatechin-3-o-gallate, l-epicatechin gallate, =--epicatechin-3-gallate, =--epicatechingallate, epicatechin monogallate, --epicatechin 3-o-gallate, teatannin, epicatechol, gallate | |
| C1C(C(OC2=CC(=CC(=C21)O)O)C3=CC(=C(C=C3)O)O)OC(=O)C4=CC(=C(C(=C4)O)O)O | |
| 442.376 | |
| CHEBI:70255 | |
| ≥98.0% (HPLC) | |
| (-)-Epicatechin Gallate |
Safety and Handling
RTECSNumber : DH9030000
TSCA : No
Recommended Storage : Refrigerator