missing translation for 'onlineSavingsMsg'
Learn More

(-)-Epicatechin Gallate 98.0+%, TCI America™

Supplier:  TCI America E0890100MG

 View more versions of this product

Catalog No. E0890100MG

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

(-)-Epicatechin Gallate
255°C
MFCD00075936
=--epicatechin gallate, epicatechin gallate, =--epicatechin-3-o-gallate, l-epicatechin gallate, =--epicatechin-3-gallate, =--epicatechingallate, epicatechin monogallate, --epicatechin 3-o-gallate, teatannin, epicatechol, gallate
C1C(C(OC2=CC(=CC(=C21)O)O)C3=CC(=C(C=C3)O)O)OC(=O)C4=CC(=C(C(=C4)O)O)O
442.376
CHEBI:70255
≥98.0% (HPLC)
1257-08-5
C22H18O10
100 mg
LSHVYAFMTMFKBA-TZIWHRDSSA-N
[(2R,3R)-2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-3,4-dihydro-2H-chromen-3-yl] 3,4,5-trihydroxybenzoate
107905
442.38
Crystalline Powder

Chemical Identifiers

1257-08-5
442.376
LSHVYAFMTMFKBA-TZIWHRDSSA-N
107905
[(2R,3R)-2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-3,4-dihydro-2H-chromen-3-yl] 3,4,5-trihydroxybenzoate
C22H18O10
MFCD00075936
=--epicatechin gallate, epicatechin gallate, =--epicatechin-3-o-gallate, l-epicatechin gallate, =--epicatechin-3-gallate, =--epicatechingallate, epicatechin monogallate, --epicatechin 3-o-gallate, teatannin, epicatechol, gallate
CHEBI:70255
C1C(C(OC2=CC(=CC(=C21)O)O)C3=CC(=C(C=C3)O)O)OC(=O)C4=CC(=C(C(=C4)O)O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : DH9030000

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions