missing translation for 'onlineSavingsMsg'
Learn More

(-)-Epicatechin, ≥97% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014305
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014305 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014305 Supplier Merck Emd Millipore Supplier No. 6809710MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

(−)-Epicatechin is a monomeric catechin and a polyphenolic component, belonging to the class of flavan-3-ols. It is majorly found in apples, apple products and green tea.

Featured Industries: Food and Beverages

Specifications

CAS 490-46-0
Shipping Condition Wet Ice
Physical Form Neat
Linear Formula C15H14O6
MDL Number MFCD00075648
Melting Point 240°C
Synonym (-)-cis-3,3′,4′,5,7-Pentahydroxyflavane; (2 R,3 R)-2-(3,4-Dihydroxyphenyl)-3,4-dihydro-1(2 H)-benzopyran-3,5,7-triol
Type Analytical Standard
Quantity 10 mg
Formula Weight 290.27
Percent Purity ≥97% (HPLC)
Grade Analytical Standard
Show More Show Less