missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ephrin B2 Recombinant Rabbit Monoclonal Antibody (23GB5270)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA552827
Description
Ephrin B2 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot
This gene encodes a member of the ephrin (EPH) family. The ephrins and EPH-related receptors comprise the largest subfamily of receptor protein-tyrosine kinases and have been implicated in mediating developmental events, especially in the nervous system and in erythropoiesis. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. This gene encodes an EFNB class ephrin which binds to the EPHB4 and EPHA3 receptors.Chemical Identifiers
| Ephrin B2 | |
| 0.8 mg/mL | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P52799, P52800 | |
| 13642, 1948, 306636 | |
| A synthesized peptide derived from human Ephrin B2 (50-100AA). | |
| Antibody |
| 23GB5270 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| Efnb2 | |
| EFNB2; efnb2a; Elf2; ELF-2; EPH-related receptor tyrosine kinase ligand 5; ephrin B2; ephrin B2a; ephrin-B2; Ephrin-B2a; Epl5; EPLG5; HTK ligand; Htkl; HTK-L; id:ibd5072; Lerk5; LERK-5; ligand of eph-related kinase 5; MGC126226; MGC126227; MGC126228; NLERK-1 | |
| Efnb2 | |
| Primary | |
| Affinity chromatography |
Specifications
| Ephrin B2 | |
| Recombinant Monoclonal | |
| 0.8 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P52799, P52800 | |
| Efnb2 | |
| A synthesized peptide derived from human Ephrin B2 (50-100AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5270 | |
| Unconjugated | |
| Efnb2 | |
| EFNB2; efnb2a; Elf2; ELF-2; EPH-related receptor tyrosine kinase ligand 5; ephrin B2; ephrin B2a; ephrin-B2; Ephrin-B2a; Epl5; EPLG5; HTK ligand; Htkl; HTK-L; id:ibd5072; Lerk5; LERK-5; ligand of eph-related kinase 5; MGC126226; MGC126227; MGC126228; NLERK-1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 13642, 1948, 306636 | |
| -20°C | |
| Liquid |