missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ephrin A1 Recombinant Rabbit Monoclonal Antibody (23GB5265)
GREENER_CHOICE

Catalog No. MA553878
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553878 100 μL
MA553879 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553878 Supplier Invitrogen™ Supplier No. MA553878
Only null left

Rabbit Recombinant Monoclonal Antibody

Ephrin A1 Recombinant Monoclonal Antibody for Western Blot

This gene encodes a member of the ephrin family. The ephrins and EPH-related receptors comprise the largest subfamily of receptor protein-tyrosine kinases and have been implicated in mediating developmental events, especially in the nervous system and in erythropoiesis. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B class, which are transmembrane proteins. This gene encodes an EFNA class ephrin which binds to the EPHA2, EPHA4, EPHA5, EPHA6, and EPHA7 receptors. Two transcript variants that encode different isoforms were identified through sequence analysis.

Specifications

Antigen Ephrin A1
Applications Western Blot
Classification Recombinant Monoclonal
Clone 23GB5265
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene EFNA1
Gene Accession No. P20827
Gene Alias AI325262; B61; ECKLG; EFL1; Efna1; Epgl1; EPH-related receptor tyrosine kinase ligand 1; ephrin A1; ephrin-A1; Ephrin-A1, secreted form; Epl1; EPLG1; Immediate early response protein B61; Lerk1; LERK-1; ligand of eph-related kinase 1; TNF alpha-induced protein 4; TNFAIP4; tumor necrosis factor alpha-induced protein 4; tumor necrosis factor, alpha-induced protein 4
Gene Symbols EFNA1
Host Species Rabbit
Immunogen A synthesized peptide derived from human Ephrin A1 (1-50AA).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1942
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less