missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EphB1 Recombinant Rabbit Monoclonal Antibody (23GB5335)

Catalog No. MA553889
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553889 20 μL
MA553888 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553889 Supplier Invitrogen™ Supplier No. MA553889
Only null left

Rabbit Recombinant Monoclonal Antibody

EphB1 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

EPHB1 is a receptor tyrosine kinase that belongs to the ephrin receptor family. Members of the Eph family of kinases play important roles in diverse biological processes including nervous system development, angiogenesis and neural synapsis formation and maturation. EphB1 and other ephB family members are type 1 membrane spanning proteins, comprised of immunoglobulin, fibronectin type III, and cysteine rich subdomains in the ecto domain, and the single uninterrupted cytoplasmic tyrosine kinase domain upstream of a carboxyterminal sterile alpha motif (SAM) domain. ephB family proteins bind ephrins of the B class, ligands that are also transmembrane spanning proteins capable of transmitting signals. ephB1 is expressed predominantly in developing neural structures in embryos, and in vascular epithelium of kidney, and other tissues. Upon binding to alternatively oligomerized ephrin B1, ephB1 signals regulation of cell attachment and cellcell assembly. Members of this protein family are implicated in neuronal and vascular cell targeting.

Specifications

Antigen EphB1
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB5335
Concentration 0.65 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene EPHB1
Gene Accession No. P54762, Q8CBF3
Gene Alias 9330129L11; AW488255; C130099E04Rik; Cek6; EK6; ELK; Elkh; ELK-related protein tyrosine kinase; ENSMUSG00000074119; Eph receptor B1; Eph receptor B2 (ELK-related protein tyrosine kinase); eph tyrosine kinase 2; EPHB1; Ephb2; EPH-like kinase 6; Ephrin type-B receptor 1; EPHT2; Epth2; Erk; Hek6; NET; Neuronally-expressed EPH-related tyrosine kinase; soluble EPHB1 variant 1; tyrosine-protein kinase receptor EPH-2
Gene Symbols EPHB1
Host Species Rabbit
Immunogen A synthesized peptide derived from human Eph receptor B1 (575-625AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2047, 270190
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less