missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EPB42 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5150056
Description
EPB42 Polyclonal Antibody for Western Blot, ICC/IF, IHC
Erythrocyte membrane protein band 4.2 is an ATP-binding protein which may regulate the association of protein 3 with ankyrin. It probably has a role in erythrocyte shape and mechanical property regulation. Mutations in the EPB42 gene are associated with recessive spherocytic elliptocytosis and recessively transmitted hereditary hemolytic anemia. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Chemical Identifiers
| EPB42 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P16452 | |
| 2038 | |
| Recombinant Erythrocyte Membrane Protein Band 4.2 (EPB42) (Leu475-Ala686). | |
| Antibody |
| 0.5 mg/mL | |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| EPB42 | |
| BEP42; E42P; Epb4.2; Epb42; Erythrocyte membrane protein band 4.2; erythrocyte protein 4.2; erythrocyte protein band 4.2; erythrocyte surface protein band 4.2; membrane skeletal protein; P4.2; PA; SPH5 | |
| EPB42 | |
| Primary | |
| Protein A |
Specifications
| EPB42 | |
| Polyclonal | |
| Unconjugated | |
| EPB42 | |
| BEP42; E42P; Epb4.2; Epb42; Erythrocyte membrane protein band 4.2; erythrocyte protein 4.2; erythrocyte protein band 4.2; erythrocyte surface protein band 4.2; membrane skeletal protein; P4.2; PA; SPH5 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 2038 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P16452 | |
| EPB42 | |
| Recombinant Erythrocyte Membrane Protein Band 4.2 (EPB42) (Leu475-Ala686). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody |