missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epalrestat 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 224°C |
| Quantity | 1 g |
| UN Number | 2811 |
| Formula Weight | 319.39 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Epalrestat |
Safety and Handling
| RTECSNumber | XJ5131855 |
| TSCA | No |