missing translation for 'onlineSavingsMsg'
Learn More

EPA 8040A Phenol Calibration Mix Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111022558
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022558 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022558 Supplier Merck Emd Millipore Supplier No. 47899

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Packaging Ampule
Flash Point 12°C (Closed Cup)
UN Number UN1219 - Class 3 - PG 2 - Isopropanol, solution
Type Organic Pollutant Standard
Quantity 1 mL
Grade Analytical Standard