missing translation for 'onlineSavingsMsg'
Learn More

EPA 8010 Surrogate Mix, 2,000 g/mL Each Component in Methanol, Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111017190
Change view
Click to view available options
Quantity:
4 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017190 4 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017190 Supplier Merck Emd Millipore Supplier No. 482344X1ML
Only null left

Specifications

Packaging Ampule
Concentration 2,000 μg/mL each component in methanol
Type Organic Pollutant Standard
Quantity 4 x 1 mL
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) Bromochloromethane; 2-Bromo-1-chloropropane; 1,4-Dichlorobutane