missing translation for 'onlineSavingsMsg'
Learn More

EPA 610-N PAH Kit Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111022551
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022551 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022551 Supplier Merck Emd Millipore Supplier No. 47351

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The EPA 610-N PAH Kit can be utilized for identifying and quantifying polynuclear aromatic hydrocarbons (PAHs) in municipal and industrial wastewater. The pre-requisite of EPA 610 method is the use of gas chromatography with a selective detector, or for gas chromatography/mass spectrometry.

  • The analyst may be better equipped to choose between GC and HPLC
  • Packed GC columns are described in the 610 method, but chromatographic conditions can be changed (e.g., capillary columns may be used) if the changes do not make the method less accurate or less precise than the standard method
  • EPA 610-N PAH kit contains 16 individually packaged polynuclear aromatic hydrocarbon neats for customizing solutions to meet the unique needs of the laboratory
  • Each PAH has been tested by GC for purity

Specifications

Flash Point Not Applicable
Physical Form Neat
UN Number UN 3316 9
Type Organic Pollutant Standard
Quantity 1 Kit
Grade Analytical Standard