missing translation for 'onlineSavingsMsg'
Learn More

EPA 547 Glyphosate Solution, Certified Reference Material, 1,000 μg/mL in H2O, MilliporeSigma™ Supelco™

Catalog No. 111001798
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111001798 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111001798 Supplier Merck Emd Millipore Supplier No. 44690U

missing translation for 'validMainframeMsg'

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Specifications

Packaging Ampule
Product Line TraceCERT
Concentration 1000 μg/mL in H2O
Synonym Glyphosate solution; N-(Phosphonomethyl)glycine
Type Organic Pollutant Standard
Quantity 1 mL
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC