missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPA 524.2 Fortification Solution, Certified Reference Material, 2,000 μg/mL Each Component in Methanol, MilliporeSigma™ Supelco™
Specifications
Specifications
| Packaging | Ampule |
| Product Line | TraceCERT |
| Concentration | 2,000 μg/mL each component in methanol |
| Type | Organic Pollutant Standard |
| Quantity | 1 mL |
| Grade | TraceCERT™ Analytical Standard |
| Concentration or Composition (by Analyte or Components) | 4-Bromofluorobenzene; 1,2-Dichlorobenzene-d4; Fluorobenzene |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC