missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPA 521 Nitrosamine Mix, Certified Reference Material, 2,000 μg/mL Each Component in Dichloromethane, MilliporeSigma™ Supelco™
Specifications
Specifications
| Packaging | Ampule |
| Product Line | TraceCERT |
| Concentration | 2,000 μg/mL each component in dichloromethane |
| Type | Organic Pollutant Standard |
| Quantity | 1 mL |
| Grade | TraceCERT™ Analytical Standard |
| Concentration or Composition (by Analyte or Components) | N-Nitrosodimethylamine; N-Nitrosodibutylamine; N-Nitrosodi-n-propylamine; N-Nitrosomethylethylamine; N-Nitrosodiethylamine; 1-Nitrosopyrrolidine; 1-Nitrosopiperidine |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC