missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EP Reference Solution Y7, Yellow Reference Solution, Ricca Chemical
Specifications
Specifications
| Chemical Name or Material | EP Reference Solution Y7 |
| Color | Colorless |
| Name Note | Yellow Reference Solution |
| CAS | 10025-77-1, 7647-01-0, 7732-18-5, 7791-13-1 |
| Grade | Laboratory |
| Odor | Odorless |
| Packaging | Amber Glass Bottle |
| Solvent or Matrix | Aqueous Solution |
| pH | Acidic |
| Quantity | 120 mL |
| Show More |