missing translation for 'onlineSavingsMsg'
Learn More

EP Reference Solution Y3, Yellow Reference Solution, Ricca Chemical

Catalog No. 288334
Change view
Click to view available options
Quantity:
120 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
288334 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 288334 Supplier Ricca Chemical Company Supplier No. 2883.34

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Yellow Standard Solution
Packaging Amber Glass Bottle
CAS 7647-01-0, 10025-77-1, 7791-13-1, 7732-18-5
Physical Form Liquid
Color Yellow to Red
pH Acidic
Melting Point 0.0°C
Boiling Point 100°C
For Use With (Equipment) Water Analyzer
Solubility Miscible with water
Solution Type Calibration Solution
Type Color Reference Standard
Quantity 120 mL
Grade Laboratory
Show More Show Less