missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Eotaxin ProQuantum Immunoassay Kit
ELISA
$990.00 - $4150.00
Specifications
| Accession Number | P51671 |
|---|---|
| Assay Range | 0.064 to 5000 pg/mL |
| Assay Sensitivity | <0.05 pg/mL |
| Conjugate | FAM™ |
| Detection Method | Taqman™ Real-time PCR |
Description
This gene is one of several Cys-Cys (CC) cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The cytokine encoded by this gene displays chemotactic activity for eosinophils, but not mononuclear cells or neutrophils. This eosinophil specific chemokine assumed to be involved in eosinophilic inflammatory diseases such as atopic dermatitis, allergic rhinitis, asthma and parasitic infections.Specifications
| P51671 | |
| <0.05 pg/mL | |
| Taqman™ Real-time PCR | |
| ProQuantum | |
| Human | |
| qPCR Instruments | |
| Scya11 | |
| Assay Dilution Buffer, Antibody-Conjugate A, Antibody-Conjugate B, Antibody-Conjugate Dilution Buffer, Standard, Master Mix, Ligase | |
| RUO | |
| Frozen at -20°C | |
| 30 mins |
| 0.064 to 5000 pg/mL | |
| FAM™ | |
| Proximity-based Immunoassay | |
| Plasma, Serum, Supernatant | |
| Protein Quantitation | |
| 6356 | |
| 1 hr. | |
| FAM™ | |
| Plasma 2-5 μL, Serum 2-5 μL, Supernatant 2-5 μL | |
| Eotaxin | |
| 2 hrs |