missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Eosin Yellowish

Catalog No. p-7027634
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2453518 50 g
AAB2453530 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2453518 Supplier Thermo Scientific Chemicals Supplier No. B2453518
Only null left

CAS: 17372-87-1 | C20H12Br4Na2O8 | 745.904 g/mol

Eosin Yellowish noted to reduce the affinity of Mg2+-ATPase for ATP and increase the enzyme affinity for Mg2+ at concentrations of 10-50 M. At higher concentrations near 100 ?M the affinity for Mg2+-ATPase for the ion-activator has been shown to be reduced. This compound has also been reported to be a photosensitizer that works as a molecular photoelectrode via catalyzation of the visible-light-driven electron-transfer reaction. Eosin Y is also used to stain structures such as muscle, cytoplasm, and collagen. This agent also enhances fluorescent quenching abilities by forming a complex with polyvinulpyrrolidone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Eosin Yellowish noted to reduce the affinity of Mg2+-ATPase for ATP and increase the enzyme affinity for Mg2+ at concentrations of 10-50 μM. At higher concentrations near 100 μM the affinity for Mg2+-ATPase for the ion-activator has been shown to be reduced. This compound has also been reported to be a photosensitizer that works as a molecular photoelectrode via catalyzation of the visible-light-driven electron-transfer reaction. Eosin Y is also used to stain structures such as muscle, cytoplasm, and collagen. This agent also enhances fluorescent quenching abilities by forming a complex with polyvinulpyrrolidone.

Solubility
Soluble in water, fairly soluble in alcohol, insoluble in ether

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 17372-87-1
Molecular Formula C20H12Br4Na2O8
Molecular Weight (g/mol) 745.904
MDL Number MFCD00133309
InChI Key MASXMTNVNASWNH-UHFFFAOYSA-L
Synonym eosin yellowish, eosine, bromo acid, eosine yellowish, eosine g, bromoeosine, c.i. acid red 87, sodium eosine, sodium eosinate, eosine salt free
PubChem CID 91886399
IUPAC Name disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate
SMILES C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+]

Specifications

Odor Odorless
Quantity 50 g
Beilstein 3586809
Merck Index 14,3603
Solubility Information Soluble in water, fairly soluble in alcohol, insoluble in ether
Formula Weight 691.86
Chemical Name or Material Eosin Yellowish
Hazard Category H319
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 241-409-6
RTECSNumber LM5850000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only