missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EOGT Recombinant Rabbit Monoclonal Antibody (23GB3585)

Catalog No. MA553415
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553415 20 μL
MA553414 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553415 Supplier Invitrogen™ Supplier No. MA553415
Only null left

Rabbit Recombinant Monoclonal Antibody

EOGT Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

The exact function of C3orf64 remains unknown.

Specifications

Antigen EOGT
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB3585
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Eogt
Gene Accession No. Q5NDL0, Q5NDL2, Q8BYW9
Gene Alias A130022J15Rik; Aer61; AER61 glycosyltransferase; AI447490; AOS4; AW214473; AW259391; C3orf64; EGF domain specific O-linked N-acetylglucosamine transferase; EGF domain-specific O-linked N-acetylglucosamine (GlcNAc) transferase; EGF domain-specific O-linked N-acetylglucosamine transferase; EGF-O-GlcNAc transferase; EOGT; EOGT1; Extracellular O-linked N-acetylglucosamine transferase; glycosyltransferase Aer61
Gene Symbols Eogt
Host Species Rabbit
Immunogen A synthetic peptide of human AER61 (1-150AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 101351, 285203, 494219
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less