missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Enzastaurin, 99%
$284.56 - $937.78
Chemical Identifiers
| CAS | 170364-57-5 |
|---|---|
| Molecular Formula | C32H29N5O2 |
| Synonym | LY317615; 3-(1-Methyl-1H-indol-3-yl)-4-[1-[1-(2-pyridinylmethyl)-4-piperidinyl]-1H-indol-3-yl]-1H-pyrrole-2,5-dione |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAJ67480MA
|
Thermo Scientific Chemicals
J67480MA |
10 mg |
Each for $284.56
|
|
|||||
|
AAJ67480MB
|
Thermo Scientific Chemicals
J67480MB |
25 mg |
Each for $526.16
|
|
|||||
|
AAJ67480MF
|
Thermo Scientific Chemicals
J67480MF |
50 mg |
Each for $937.78
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 170364-57-5 | |
| C32H29N5O2 | |
| LY317615; 3-(1-Methyl-1H-indol-3-yl)-4-[1-[1-(2-pyridinylmethyl)-4-piperidinyl]-1H-indol-3-yl]-1H-pyrrole-2,5-dione |
Specifications
| Red to orange | |
| 170364-57-5 | |
| 515.60 | |
| 249°C to 261°C | |
| Powder | |
| AXRCEOKUDYDWLF-UHFFFAOYSA-N | |
| 3-(1-methyl-1H-indol-3-yl)-4-(1-{1-[(pyridin-2-yl)methyl]piperidin-4-yl}-1H-indol-3-yl)-2,5-dihydro-1H-pyrrole-2,5-dione | |
| 99% |
| -30°C to -10°C | |
| C32H29N5O2 | |
| LY317615; 3-(1-Methyl-1H-indol-3-yl)-4-[1-[1-(2-pyridinylmethyl)-4-piperidinyl]-1H-indol-3-yl]-1H-pyrrole-2,5-dione | |
| Enzastaurin, 99% | |
| MFCD11040980 | |
| CN1C=C(C2=CC=CC=C12)C1=C(C(=O)NC1=O)C1=CN(C2CCN(CC3=CC=CC=N3)CC2)C2=CC=CC=C12 | |
| 515.62 |
Safety and Handling
Recommended Storage : Store at -20°C
RUO – Research Use Only