missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Enzastaurin, 99%

Catalog No. AAJ67480MA
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67480MA 10 mg
AAJ67480MB 25 mg
AAJ67480MF 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ67480MA Supplier Thermo Scientific Chemicals Supplier No. J67480MA

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 170364-57-5
Molecular Formula C32H29N5O2
Synonym LY317615; 3-(1-Methyl-1H-indol-3-yl)-4-[1-[1-(2-pyridinylmethyl)-4-piperidinyl]-1H-indol-3-yl]-1H-pyrrole-2,5-dione

Specifications

Quantity 10 mg
Description Red to orange
Content And Storage -30°C to -10°C
Formula Weight 515.60
Melting Point 249°C to 261°C
Product Type Enzastaurin, 99%
Form Powder
MDL Number MFCD11040980
InChI Key AXRCEOKUDYDWLF-UHFFFAOYSA-N
SMILES CN1C=C(C2=CC=CC=C12)C1=C(C(=O)NC1=O)C1=CN(C2CCN(CC3=CC=CC=N3)CC2)C2=CC=CC=C12
IUPAC Name 3-(1-methyl-1H-indol-3-yl)-4-(1-{1-[(pyridin-2-yl)methyl]piperidin-4-yl}-1H-indol-3-yl)-2,5-dihydro-1H-pyrrole-2,5-dione
Molecular Weight (g/mol) 515.62
Assay 99%
Show More Show Less
Recommended Storage Store at -20°C

RUO – Research Use Only