missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Entris™ II Advanced Precision Balances
GREENER_CHOICE

Catalog No. 14558898
Change view
Click to view available options
Capacity:
220 g
320 g
420 g
620 g
820 g
1.2 kg
2.2 kg
3.2 kg
4.2 kg
5.2 kg
6.2 kg
8.2 kg
10.2 kg
12.2 kg
Readability:
1 mg
10 mg
100 mg
Legal for Trade:
Class I
Class II
Class III
Calibration:
Internal
31 product options available for selection
Product selection table with 31 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity Readability Legal for Trade Calibration
14558898 320 g 1 mg Class I -
14558893 220 g 1 mg Class I Internal
14558892 220 g 1 mg Class I -
14558899 320 g 1 mg Class I Internal
14558904 420 g 1 mg Class I Internal
14558903 420 g 1 mg Class I -
14558910 620 g 1 mg Class I Internal
14558909 620 g 1 mg Class I -
14558916 820 g 10 mg Class II Internal
14558915 820 g 10 mg Class II -
14558883 1.2 kg 1 mg - Internal
14558882 1.2 kg 10 mg Class II Internal
14558881 1.2 kg 10 mg Class II -
14558891 2.2 kg 10 mg Class II Internal
14558890 2.2 kg 10 mg Class II -
14558889 2.2 kg 100 mg Class III Internal
14558888 2.2 kg 100 mg Class III -
14558897 3.2 kg 10 mg Class II Internal
14558896 3.2 kg 10 mg Class II -
14558902 4.2 kg 10 mg Class II Internal
14558901 4.2 kg 10 mg Class II -
14558906 5.2 kg 100 mg Class III Internal
14558905 5.2 kg 100 mg Class III -
14558908 6.2 kg 10 mg Class II Internal
14558907 6.2 kg 10 mg Class II -
14558914 8.2 kg 100 mg Class III Internal
14558913 8.2 kg 100 mg Class III -
14558880 10.2 kg 100 mg Class III Internal
14558879 10.2 kg 100 mg Class III -
14558885 12.2 kg 100 mg Class III Internal
14558884 12.2 kg 100 mg Class III -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 31 options available.
31 options
Catalog No. 14558898 Supplier Sartorius Supplier No. BCA3231S
Only null left

Over 38 different models offer various benefits such as real-time level support, integrated protection systems, CalAuditTrail, a graphic touch display, and 13 built-in application programs

  • Entris™ II Advanced balances - Quality, Value, and Consistency
  • Accurate weighing results that can be trusted time after time
  • Simplifies balance leveling with built-in, real-time level support
  • Increases reliability of weighing results by using integrated protection systems
  • Controlled access to balance settings with user management
  • Gapless documentation thanks to CalAuditTrail

Specifications

Certifications/Compliance CE Certificate, CSA for Canada
Connection Type D-sub connector, 9-pin; USB UTL; USB Device
Description Without Internal Calibration
Capacity 320 g
Display Type Intuitive Graphic Touch Technology
Readability 1 mg
Material Glass, PBT, Stainless Steel
Model Electronic Precision Balance
Stabilization Time ≤1.0 seconds
Type Balance
Voltage 100/240 V
Amperage 0.2 A
Frequency 50/60 Hz
Legal for Trade Class I
Show More Show Less