missing translation for 'onlineSavingsMsg'
Learn More

(±)-Enterolactone, ≥95.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015546
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015546 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015546 Supplier Merck Emd Millipore Supplier No. 4236310MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 78473-71-9
Linear Formula C18H18O4
MDL Number MFCD01861506
Synonym Enterolactone; trans-alpha,beta-Bis(3-hydroxybenzyl)butyrolactone
Type Analytical Standard
Quantity 10 mg
Formula Weight 298.33
Percent Purity ≥95.0% (HPLC)
Grade Analytical Standard