missing translation for 'onlineSavingsMsg'
Learn More

Enrofloxacin Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015921
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015921 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015921 Supplier Merck Emd Millipore Supplier No. PHR1513500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Enrofloxacin is classified under the second generation quinolone antibiotics, known as ′fluoroquinolones′. It is a component of several medicines used to treat urinary tract infections, infections of the gastrointestinal tract, respiratory tract infections, infections of the skin structure as well as surgical prophylaxis. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 93106-60-6
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C19H22 FN3O3
MDL Number MFCD00792463
Synonym Enrofloxacin; Baytril
Type Certified Reference Material
Quantity 500 mg
Formula Weight 359.39
Grade Certified Reference Material