missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ENOX2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117085
Description
Positive Samples: MCF7, THP-1, SW480, Jurkat, SKOV3, Mouse liver, Mouse kidney, Mouse ovary, Rat liver Immunogen sequence: AEALKEENDS LRWQLDAYRN EVELLKQEQG KVHREDDPNK EQQLKLLQQA LQGMQQHLLK VQEEYKKKEA ELEKLKDDKL QVEKMLENLK EKESCASRLC ASNQDSEYPL EKTMNSSPIK SEREALLVGI ISTFLHVHPF GASIEYICSY LHRLDNKICT SDVECLMGRL QHTFKQEMTG VGASLEKRWK FCGFEGLKLT.
This gene is a tumor-specific member of the ECTO-NOX family of genes that encode cell surface NADH oxidases. The encoded protein has two enzymatic activities: catalysis of hydroquinone or NADH oxidation, and protein disulfide interchange. The protein also displays prion-like properties. Alternative splicing results in multiple transcript variants.Chemical Identifiers
| ENOX2 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| Enox2 | |
| APK1; APK1 antigen; Cova1; cytosolic ovarian carcinoma antigen 1; Ecto-NOX disulfide-thiol exchanger 2; enox2; Hydroquinone [NADH] oxidase; Protein disulfide-thiol oxidoreductase; RGD1563643; tNOX; tumor-associated hydroquinone oxidase; zmp:0000001197 | |
| Enox2 | |
| Primary | |
| Affinity chromatography |
| 3.12 mg/mL | |
| ELISA, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q16206, Q8R0Z2 | |
| 10495, 209224, 302817 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 411-610 of human ENOX2 (NP_8721141). | |
| Antibody |
Specifications
| ENOX2 | |
| Polyclonal | |
| Unconjugated | |
| Enox2 | |
| APK1; APK1 antigen; Cova1; cytosolic ovarian carcinoma antigen 1; Ecto-NOX disulfide-thiol exchanger 2; enox2; Hydroquinone [NADH] oxidase; Protein disulfide-thiol oxidoreductase; RGD1563643; tNOX; tumor-associated hydroquinone oxidase; zmp:0000001197 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10495, 209224, 302817 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 3.12 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q16206, Q8R0Z2 | |
| Enox2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 411-610 of human ENOX2 (NP_8721141). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |