missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Endothall

A moderately potent inhibitor of protein phosphatase 2A

$713.79

Chemical Identifiers

CAS 145-73-3
Molecular Formula C8H10O5, C8H8Na2O5
Synonym 7-Oxabicyclo[2.2.1]heptane-2,3-dicarboxylic acid
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ60948MC
View Documents Promotion Details
Thermo Scientific Chemicals
J60948MC
100 mg
Each for $713.79
Only null left
 
Description

Description

Endothall is used as an inhibitor of PP2A phosphatase activity. It is also used as a defoliant. It is used as a cell adhesion inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

145-73-3
C8H10O5, C8H8Na2O5
7-Oxabicyclo[2.2.1]heptane-2,3-dicarboxylic acid

Specifications

White
A moderately potent inhibitor of protein phosphatase 2A
C8H10O5, C8H8Na2O5
7-Oxabicyclo[2.2.1]heptane-2,3-dicarboxylic acid
Powder
[Na+].[Na+].[O-]C(=O)C1C2CCC(O2)C1C([O-])=O
230.13
2°C to 8°C
145-73-3
186.20g/mol
Signal Transduction Reagent
XRHVZWWRFMCBAZ-UHFFFAOYNA-L
disodium 7-oxabicyclo[2.2.1]heptane-2,3-dicarboxylate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 205-660-5

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only