accessibility menu, dialog, popup

UserName

CAS RN 145-73-3

IUPAC Name: disodium 7-oxabicyclo[2.2.1]heptane-2,3-dicarboxylate
Synonyms: disodium 7-oxabicyclo[2.2.1]heptane-2,3-dicarboxylate
Molecular Weight (g/mol): 230.13
Molecular Formula: C8H8Na2O5
InChi Key: XRHVZWWRFMCBAZ-UHFFFAOYNA-L
SMILES: [Na+].[Na+].[O-]C(=O)C1C2CCC(O2)C1C([O-])=O
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results