missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ENDOGL1 Recombinant Rabbit Monoclonal Antibody (23GB1370)

Rabbit Recombinant Monoclonal Antibody

Supplier:  Invitrogen™ MA553999

 View more versions of this product

Catalog No. MA553999


Only null left

Description

Description

ENDOGL1 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

This gene encodes an endo/exonuclease with 5'-3' exonuclease activity. The encoded enzyme catalyzes the hydrolysis of ester linkages at the 5' end of a nucleic acid chain. This enzyme is localized to the mitochondria and may play a role in programmed cell death. Alternatively spliced transcript variants have been described. A pseudogene exists on chromosome 18.
Specifications

Chemical Identifiers

ENDOGL1
0.8 mg/mL
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q9Y2C4
9941
A synthesized peptide derived from human ENDOGL1/ENGL (350-368AA).
Antibody
23GB1370
-20°C
Recombinant Monoclonal
Liquid
IgG
Human
EXOG
AW557704; Endo G-like 1; endo/exonuclease (5'-3'), endonuclease G-like; endo/exonuclease (5-3), endonuclease G-like; ENDOGL1; Endogl2; endonuclease G-like 1; endonuclease G-like 2; ENGL; Engla; ENGL-a; ENGLB; ENGL-b; exo/endonuclease G; EXOG; Nuclease EXOG, mitochondrial; nuclease EXOG, mitochondrial-like; Nuclease EXOG, mitochondrial-like protein
EXOG
Primary
Affinity chromatography

Specifications

ENDOGL1
Recombinant Monoclonal
0.8 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q9Y2C4
EXOG
A synthesized peptide derived from human ENDOGL1/ENGL (350-368AA).
20 μL
Primary
Human
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
23GB1370
Unconjugated
EXOG
AW557704; Endo G-like 1; endo/exonuclease (5'-3'), endonuclease G-like; endo/exonuclease (5-3), endonuclease G-like; ENDOGL1; Endogl2; endonuclease G-like 1; endonuclease G-like 2; ENGL; Engla; ENGL-a; ENGLB; ENGL-b; exo/endonuclease G; EXOG; Nuclease EXOG, mitochondrial; nuclease EXOG, mitochondrial-like; Nuclease EXOG, mitochondrial-like protein
Rabbit
Affinity chromatography
RUO
9941
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions