missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ENDOGL1 Recombinant Rabbit Monoclonal Antibody (23GB1370)

Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA553998
Description
ENDOGL1 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot
This gene encodes an endo/exonuclease with 5'-3' exonuclease activity. The encoded enzyme catalyzes the hydrolysis of ester linkages at the 5' end of a nucleic acid chain. This enzyme is localized to the mitochondria and may play a role in programmed cell death. Alternatively spliced transcript variants have been described. A pseudogene exists on chromosome 18.Chemical Identifiers
| ENDOGL1 | |
| 0.8 mg/mL | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9Y2C4 | |
| 9941 | |
| A synthesized peptide derived from human ENDOGL1/ENGL (350-368AA). | |
| Antibody |
| 23GB1370 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| EXOG | |
| AW557704; Endo G-like 1; endo/exonuclease (5'-3'), endonuclease G-like; endo/exonuclease (5-3), endonuclease G-like; ENDOGL1; Endogl2; endonuclease G-like 1; endonuclease G-like 2; ENGL; Engla; ENGL-a; ENGLB; ENGL-b; exo/endonuclease G; EXOG; Nuclease EXOG, mitochondrial; nuclease EXOG, mitochondrial-like; Nuclease EXOG, mitochondrial-like protein | |
| EXOG | |
| Primary | |
| Affinity chromatography |
Specifications
| ENDOGL1 | |
| Recombinant Monoclonal | |
| 0.8 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9Y2C4 | |
| EXOG | |
| A synthesized peptide derived from human ENDOGL1/ENGL (350-368AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB1370 | |
| Unconjugated | |
| EXOG | |
| AW557704; Endo G-like 1; endo/exonuclease (5'-3'), endonuclease G-like; endo/exonuclease (5-3), endonuclease G-like; ENDOGL1; Endogl2; endonuclease G-like 1; endonuclease G-like 2; ENGL; Engla; ENGL-a; ENGLB; ENGL-b; exo/endonuclease G; EXOG; Nuclease EXOG, mitochondrial; nuclease EXOG, mitochondrial-like; Nuclease EXOG, mitochondrial-like protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 9941 | |
| -20°C | |
| Liquid |