missing translation for 'onlineSavingsMsg'
Learn More

endo-Tetrahydrodicyclopentadiene 97.0+%, TCI America™

Catalog No. T040925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T040925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T040925G Supplier TCI America Supplier No. T040925G

Chemical Identifiers

CAS 2825-83-4
Molecular Formula C10H16
Molecular Weight (g/mol) 136.24
MDL Number MFCD00074748
InChI Key LPSXSORODABQKT-UHFFFAOYNA-N
Synonym endo-Tricyclo[5.2.1.0(2,6)]decane
PubChem CID 98044726
IUPAC Name tricyclo[5.2.1.0²,⁶]decane
SMILES C1CC2C3CCC(C3)C2C1

Specifications

Melting Point 75°C
Color White-Yellow
Boiling Point 192°C
Quantity 25 g
Formula Weight 136.24
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material endo-Tetrahydrodicyclopentadiene
EINECSNumber (4)-0655
TSCA Yes