accessibility menu, dialog, popup

UserName

CAS RN 2825-83-4

IUPAC Name: tricyclo[5.2.1.0²,⁶]decane
Synonyms: tricyclo[5.2.1.0²,⁶]decane
Molecular Weight (g/mol): 136.24
Molecular Formula: C10H16
InChi Key: LPSXSORODABQKT-UHFFFAOYNA-N
SMILES: C1CC2C3CCC(C3)C2C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results