missing translation for 'onlineSavingsMsg'
Learn More

Enalapril

Catalog No. p-7041929
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6075003 1 g
AAJ6075006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6075003 Supplier Thermo Scientific Chemicals Supplier No. J6075003

Long-acting ACE inhibitor and antihypertensive | CAS: 75847-73-3 | C20H28N2O5 | 376.453 g/mol

Enalapril is used in the treatment of hypertension, diabetic nephropathy, and some types of chronic heart failure. It has been proven to protect the function of the kidneys in hypertension, heart failure, and diabetes, and may be used in the absence of hypertension for its kidney protective effects. It is widely used in chronic kidney failure.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Enalapril is used in the treatment of hypertension, diabetic nephropathy, and some types of chronic heart failure. It has been proven to protect the function of the kidneys in hypertension, heart failure, and diabetes, and may be used in the absence of hypertension for its kidney protective effects. It is widely used in chronic kidney failure.

Solubility
Soluble in water at 25mg/ml

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.

Chemical Identifiers

CAS 75847-73-3
Molecular Formula C20H28N2O5
Molecular Weight (g/mol) 376.453
MDL Number MFCD00865774
InChI Key GBXSMTUPTTWBMN-XIRDDKMYSA-N
Synonym enalapril, enalaprilum, vasotec, enalaprila, enalapril tn, unii-69pn84io1a, enalapril inn, enalapril richet, chembl578, enalaprilum inn-latin
PubChem CID 5388962
ChEBI CHEBI:4784
IUPAC Name (2S)-1-[(2S)-2-[[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino]propanoyl]pyrrolidine-2-carboxylic acid
SMILES CCOC(=O)C(CCC1=CC=CC=C1)NC(C)C(=O)N2CCCC2C(=O)O

Specifications

Melting Point 142°C to 147°C
Color White
Quantity 1 g
Merck Index 14,3567
Solubility Information Soluble in water at 25mg/ml
Formula Weight 376.45
Physical Form Powder
Chemical Name or Material Enalapril
RTECSNumber TW3665500
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only